missing translation for 'onlineSavingsMsg'
Learn More

YY1 Rabbit anti-Human, Mouse, Rat, Clone: 6J6L1, Novus Biologicals™

Product Code. 18371692
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18371692 100 μg 100µL
18365733 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18371692 Supplier Bio-Techne Supplier No. NBP315779100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

YY1 Monoclonal antibody specifically detects YY1 in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay, Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen YY1
Applications ChIP Assay, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 6J6L1
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias DELTA, Delta transcription factor, INO80SYY-1, NF-E1INO80 complex subunit S, transcriptional repressor protein YY1, UCRBP, Yin and yang 1, Yin and Yang 1 protein, YIN-YANG-1, YY1 transcription factor
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 200-300 of human YY1 (P25490).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Chromatin Research, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7528
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.