missing translation for 'onlineSavingsMsg'
Learn More

Yunaconitine, MedChemExpress

Product Code. 16488948
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16488948

Brand: Medchem Express HYN0333

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Yunaconitine(Guayewuanine B) is a highly toxic aconitum alkaloid.

Yunaconitine(Guayewuanine B) is a highly toxic aconitum alkaloid.

Specifications

Chemical Name or Material Yunaconitine
CAS 70578-24-4
Color White
Purity Grade Notes Research
Molecular Formula C35H49NO11
Quantity 10 mg
Synonym Guayewuanine B
Solubility Information DMSO : ≥ 250 mg/mL (378.93 mM)
SMILES CO[C@@H]1C23[C@@]4([H])[C@](COC)(CN(CC)C2C([C@]5(OC(C)=O)[C@]6([H])[C@@]3([H])C[C@](O)([C@@H](OC)C5)[C@@H]6OC(C7=CC=C(OC)C=C7)=O)[C@@H]4OC)[C@H](O)C1
Molecular Weight (g/mol) 659.76
Formula Weight 659.76
Percent Purity 99.36%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.