missing translation for 'onlineSavingsMsg'
Learn More

Yttrium Standard: Y @ 5 wt % in 20% HNO3

Product Code. 30318468
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30318468 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30318468 Supplier VHG Labs Supplier No. VHGYN5P1L

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Yttrium
CAS 7440-65-5
Analyte or Component Names Yttrium
Shipping Condition Room Temperature
Molecular Formula Y
Concentration 5 %
Synonym Y
InChI Formula InChI=1 S/Y
Solution Type Mixture
SMILES [Y]
Element Y
IUPAC Name yttrium
Solvent or Matrix 20% HNO3
Industry Type Industrial QC
Molecular Weight (g/mol) 88.906
Quantity 1 L
Formula Weight 88.906
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.