missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) trifluoroacetate hydrate, 99.9% (REO)

Product Code. 11495380
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11495380 50 g 50g
11485380 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11495380 Supplier Thermo Scientific Alfa Aesar Supplier No. 044597.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 304851-95-4 | C6F9O6Y | 427.95 g/mol

A fluorinated yttrium compound for proteomics research. Used in the manufacture of ceramics, glass, laser materials, phosphor. non-ferrous alloy iron and steel, and the additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 304851-95-4
Molecular Formula C6F9O6Y
Molecular Weight (g/mol) 427.95
MDL Number MFCD05663780 MFCD00192577
InChI Key NPATVYPXYMSPKD-UHFFFAOYSA-K
Synonym yttrium trifluoroacetate
PubChem CID 22836421
IUPAC Name 2,2,2-trifluoroacetic acid;yttrium;hydrate
SMILES [Y+3].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F

Specifications

Physical Form Crystalline
Quantity 50 g
Solubility Information Insoluble in water.
Formula Weight 427.95 (Anhydrous)
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Yttrium (III) trifluoroacetate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.