missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) oxide, REacton™, 99.99% (REO)

Product Code. p-7082564
Click to view available options
Quantity:
25 g
500 g
Unit Size:
25g
500g
This item is not returnable. View return policy

Product Code. 11346198

Brand: Thermo Scientific Alfa Aesar 011181.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1314-36-9 | O3Y2 | 225.81 g/mol

Yttriium(III) oxide is used to make phosphors for color TV tubes, yttrium iron garnets for use as microwave filters, high temperature superconductors, as a mineralizing agent for the formation of spinel and as a stabilizer for high-temperature service materials. It is presurcor for important inorganic compounds. It is a potential solid-state laser material which allows the efficient operation both in continuous operation and in pulsed regimes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-36-9
Molecular Formula O3Y2
Molecular Weight (g/mol) 225.81
MDL Number MFCD00011473
InChI Key RUDFQVOCFDJEEF-UHFFFAOYSA-N
IUPAC Name diyttrium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Y+3].[Y+3]

Specifications

Melting Point 2410°C
Color White
Physical Form Powder
Quantity 25 g
Linear Formula Y2O3
Merck Index 14,10107
Solubility Information Insoluble in water. Soluble in alcohol and dilute acids
Formula Weight 225.81
Percent Purity 99.99%
Odor Odorless
Sensitivity Air Sensitive
Density 5.01
Chemical Name or Material Yttrium(III) oxide, REacton™, (REO)
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.