missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) nitrate hydrate, REacton™, 99.99% (REO)

Product Code. 11376208
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
5 x 100 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11376208 5 g 5g
11306218 25 g 25g
11396208 100 g 100g
11386208 5 x 100 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11376208 Supplier Thermo Scientific Alfa Aesar Supplier No. 011187.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 140191-40-8 | N3O9Y | 274.92 g/mol

It acts as a precursor for the preparation of yttrium oxide nano particles and for nanoscale coatings of carbon composites. It is an effective yttrium source for the preparation of yttrium-based surfactant mesophases which are used as adsorbing agents and in optically functional materials. It is used in optics, ceramics, glass, electronics, and superconducting materials. It has been reported as a reusable catalyst in Bignelli reaction for the synthesis of heterocycles such as pyrimidin-2-ones. It is a powerful catalyst for the synthesis of some supramolecules such as calix[4]resorcinarenes, 1,8-dioxooctahydroxanthenes, 2-amino-4H-chromenes, and several other organic condensation products. Silica-supported yttrium nitrate hydrate acts as a heterogeneous catalyst for the preparation of 2-Amino-4H-Chromenes in water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140191-40-8
Molecular Formula N3O9Y
Molecular Weight (g/mol) 274.92
MDL Number MFCD00011469
InChI Key BXJPTTGFESFXJU-UHFFFAOYSA-N
Synonym yttrium iii nitrate hydrate, yttrium nitrate hydrate, 3no3.y.h2o, nitric acid, yttrium iii salt, nitric acid, yttrium 3++ salt, yttrium 3+ hydrate trinitrate, yttrium iii nitrate hydrate, reacton
PubChem CID 23169389
SMILES [Y+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Physical Form Crystalline Aggregates
Quantity 5 g
UN Number UN1477
Merck Index 14,10107
Solubility Information Soluble in water.
IUPAC Name yttrium(3+);trinitrate;hydrate
Formula Weight 274.92 (Anhydrous)
Percent Purity 99.99%
Grade Reacton
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Yttrium(III) nitrate hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.