missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) isopropoxide, 90+%

Product Code. 11322428
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322428 1 g 1g
11312428 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322428 Supplier Thermo Scientific Alfa Aesar Supplier No. 014477.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2172-12-5 | C9H24O3Y | 269.19 g/mol

Yttrium(III) isopropoxide is used in asymmetric catalysis applications. It acts as a catalyst for stereoselective conjugate addition reactions and ring-opening polymerizations. It serves as a precursor for the generation of reactive enolates by enantioselective protonation reactions and nanocomposites synthesis. Further, it is used in the preparation of flat hafnium(IV) oxide films. In addition to this, it is used to improve the thermal conductivity of polycrystalline aluminum nitride (AlN) ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2172-12-5
MDL Number MFCD00015641

Specifications

Melting Point 160°C
Boiling Point 200°C to 210°C (1 mmHg)(sublimation)
Molecular Formula C22H29KO4
Quantity 1 g
UN Number UN2925
Sensitivity Moisture sensitive
Solubility Information Soluble in hexane,toluene and terahydrofuran.
InChI Key JTZQCHFUGHIPDF-RYVBEKKQSA-M
SMILES CC12CCC(=O)C=C1C=CC3C2CCC4(C3CCC4(CCC(=O)[O-])O)C.[K+]
IUPAC Name potassium;3-[(10S,13R,17S)-17-hydroxy-10,13-dimethyl-3-oxo-2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthren-17-yl]propanoate
Molecular Weight (g/mol) 396.57
PubChem CID 133109093
Formula Weight 266.17
Percent Purity ≥90%
Physical Form Powder
Chemical Name or Material Yttrium(III) isopropoxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Yttrium(III) isopropoxide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H228-Flammable solid.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Yttrium(III) isopropoxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.