missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) isopropoxide, 20-25% w/w in toluene, Thermo Scientific™

Product Code. 11400470
Change view
Click to view available options
Quantity:
∼1 g
∼25 g
5 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400470 ∼25 g 25g
11410470 ∼1 g 1g
11490460 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11400470 Supplier Thermo Scientific Alfa Aesar Supplier No. 014524.ND
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2172-12-5
Molecular Formula C9H24O3Y
Molecular Weight (g/mol) 269.19
MDL Number MFCD00015641
InChI Key NREVZTYRXVBFAQ-UHFFFAOYSA-N
Synonym yttrium isopropoxide
PubChem CID 85308566
IUPAC Name propan-2-ol;yttrium
SMILES CC(C)O.CC(C)O.CC(C)O.[Y]

Specifications

Color Yellow to Brown
Density 0.955 g/mL
Odor Aromatic
Linear Formula Y[OCH(CH3)2]3
Quantity ∼25 g
UN Number UN3274
Sensitivity Air and moisture sensitive
Solubility Information Soluble in hexane,toluene,and THF.
Formula Weight 266.17
Concentration or Composition (by Analyte or Components) 20 to 25% w/w in toluene
Physical Form Liquid
Chemical Name or Material Yttrium(III) isopropoxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.