missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) hexafluoro-2,4-pentanedionate

Product Code. p-7049564
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11347028

Brand: Thermo Scientific Alfa Aesar 014717.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18911-76-7 | C15H3F18O6Y | 710.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18911-76-7
Molecular Formula C15H3F18O6Y
Molecular Weight (g/mol) 710.06
MDL Number MFCD00064764
InChI Key AJVDSGQQIVVYRX-UHFFFAOYSA-N
Synonym yttrium hexafluoroacetylacetonate, z-1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one; yttrium
PubChem CID 50930692
SMILES [Y+3].FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F

Specifications

Melting Point 166°C to 170°C
Physical Form Crystalline
Quantity 1 g
IUPAC Name yttrium(3+) tris(1,1,1,5,5,5-hexafluoro-2,4-dioxopentan-3-ide)
Formula Weight 710.1
Chemical Name or Material Yttrium(III) hexafluoro-2,4-pentanedionate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.