missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) hexafluoro-2,4-pentanedionate

Product Code. 11357028
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357028 5 g 5g
11347028 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357028 Supplier Thermo Scientific Alfa Aesar Supplier No. 014717.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18911-76-7 | C15H3F18O6Y | 710.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18911-76-7
Molecular Formula C15H3F18O6Y
Molecular Weight (g/mol) 710.06
MDL Number MFCD00064764
InChI Key AJVDSGQQIVVYRX-UHFFFAOYSA-N
Synonym yttrium hexafluoroacetylacetonate, z-1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one; yttrium
PubChem CID 50930692
SMILES [Y+3].FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F

Specifications

Melting Point 166°C to 170°C
Physical Form Crystalline
Quantity 5 g
IUPAC Name (Z)-1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one;yttrium
Formula Weight 710.1
Chemical Name or Material Yttrium(III) hexafluoro-2,4-pentanedionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.