missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) fluoride, anhydrous, REacton™, 99.9% (REO)

Product Code. 11366898
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11366898 25 g 25g
11346898 100 g 100g
11356898 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11366898 Supplier Thermo Scientific Alfa Aesar Supplier No. 013650.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13709-49-4 | F3Y | 145.90 g/mol

Yttrium(III) fluoride is used in the production of yttrium metal, glasses and ceramics. It is also used in the preparation of low-index film layers, which exhibit good transparency in the UV through infrared regions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13709-49-4
Molecular Formula F3Y
Molecular Weight (g/mol) 145.90
MDL Number MFCD00049714
InChI Key RBORBHYCVONNJH-UHFFFAOYSA-K
IUPAC Name yttrium(3+) trifluoride
SMILES [F-].[F-].[F-].[Y+3]

Specifications

Melting Point 1387°C
Physical Form Lump/Powder
Quantity 25 g
UN Number UN3288
Solubility Information Soluble in acids. Insoluble in water.
Formula Weight 145.9
Percent Purity 99.9%
Grade Reacton
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Yttrium(III) fluoride, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.