missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) chloride, ultra dry, 99.99% (REO)

Product Code. p-7050161
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11398617

Brand: Thermo Scientific Alfa Aesar 035743.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10361-92-9

Yttrium(III) chloride is used as a precursor for titanium-containing ceramics. It is used as an analytical reagent, as a catalyst and in superconductors. It is also used in the production of yttrium alkyl alkoxide complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10361-92-9
Molecular Formula Cl3Y
Molecular Weight (g/mol) 195.26
MDL Number MFCD00011470
InChI Key PCMOZDDGXKIOLL-UHFFFAOYSA-K
IUPAC Name trichloroyttrium
SMILES Cl[Y](Cl)Cl

Specifications

Melting Point 721°C
Color White to Yellow
Physical Form Powder
Quantity 10 g
Linear Formula YCl3
Merck Index 14,10107
Solubility Information Soluble in water,ethanol and pyridine.
Formula Weight 195.26
Percent Purity 99.99%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 2.67 g/mL
Assay (REO)
Chemical Name or Material Yttrium(III) chloride, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.