missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) chloride hydrate, REacton™, 99.99% (REO)

Product Code. 11346208
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
250 g
Unit Size:
100g
250g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11346208 250 g 250g
11326208 5 g 5g
11316208 25 g 25g
11336208 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11346208 Supplier Thermo Scientific Alfa Aesar Supplier No. 011185.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12741-05-8 | Cl3Y | 195.26 g/mol

Yttrium(III) chloride is used a precursor for the synthesis of optically active crystals and as a starting material for the synthesis of polymerization catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12741-05-8
Molecular Formula Cl3Y
Molecular Weight (g/mol) 195.26
MDL Number MFCD00149941
InChI Key PCMOZDDGXKIOLL-UHFFFAOYSA-K
Synonym yttrium iii chloride hydrate, yttrium chloride, hydrate, acmc-1bq0g, ksc171o8f, yttrium chloride-water 1/3/1, yttrium 3+ ion hydrate trichloride
PubChem CID 57347377
IUPAC Name yttrium(3+);trichloride;hydrate
SMILES [Cl-].[Cl-].[Cl-].[Y+3]

Specifications

Color Pink to White
Physical Form Crystalline Aggregates
Quantity 250 g
Linear Formula YCl3·xH2O
Merck Index 14,10107
Solubility Information Soluble in water,pyridine,ethanol.
Formula Weight 195.26 (Anhydrous)
Percent Purity 99.99%
Grade Reacton
Odor Odorless
Sensitivity Hygroscopic
Density 2.18g/mL
Assay (REO)
Chemical Name or Material Yttrium(III) chloride hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.