missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) bromide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

£92.20 - £350.00

Chemical Identifiers

CAS 13469-98-2
MDL Number MFCD00049710
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11358627
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035746.04
2 g
£92.20
2g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11348627
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035746.09
10 g
£350.00
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Yttrium(III) bromide is used in electroplating, as catalyst, material enhancing & medical treatments in curing colds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
Specifications

Chemical Identifiers

13469-98-2
MFCD00049710

Specifications

904°C
Powder
It is soluble in water at a concentration of 83.3g/100 mL at 30°C.
99.9%
Hygroscopic
Yttrium(III) bromide, Ultra dry
13469-98-2
MFCD00049710
328.63
Ampule
(REO)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 236-728-2

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Ampouled under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only