missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) bromide, ultra dry, 99.9% (REO)

Product Code. 11348627
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348627 10 g 10g
11358627 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348627 Supplier Thermo Scientific Alfa Aesar Supplier No. 035746.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 13469-98-2

Yttrium(III) bromide is used in electroplating, as catalyst, material enhancing & medical treatments in curing colds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13469-98-2
MDL Number MFCD00049710

Specifications

Melting Point 904°C
Physical Form Powder
Quantity 10 g
Solubility Information It is soluble in water at a concentration of 83.3g/100 mL at 30°C.
Formula Weight 328.63
Percent Purity 99.9%
Packaging Ampule
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Yttrium(III) bromide, Ultra dry

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.