missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Yttrium(III) acetate tetrahydrate, 99.9% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
In analytical chemistry.Yttrium(III) acetate tetrahydrate is used in analytical chemistry and in catalysis. It is used in the preparation of yttrium trifluoride nanoparticles, which shows antibacterial properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | (decomposition) |
Color | White |
Physical Form | Powder |
Quantity | 50 g |
Linear Formula | Y(OOCCH3)3·4H2O |
Solubility Information | Soluble in hot water. |
Formula Weight | 338.10 (266.04 Anhydrous) |
Percent Purity | 99.9% |
Density | 1.5 g/mL |
Assay | (REO) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'