missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) 2-methoxyethoxide, 5% w/v in 2-methoxyethanol, Thermo Scientific™

Product Code. 11398948
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398948 100 mL 100mL
11308958 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398948 Supplier Thermo Scientific Alfa Aesar Supplier No. 041793.AE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115668-57-0
Molecular Formula C9H21O6Y
Molecular Weight (g/mol) 314.17
MDL Number MFCD00156527,MFCD00156527
InChI Key IVXUPDRNSUCVNX-UHFFFAOYSA-N
Synonym 1-methoxyethanolate,yttrium 3+, 2-methoxyethanolate;yttrium 3+
PubChem CID 14452922
IUPAC Name 2-methoxyethanolate;yttrium(3+)
SMILES [Y+3].COCC[O-].COCC[O-].COCC[O-]

Specifications

Linear Formula Y(OCH2CH2OCH3)3
Quantity 100 mL
UN Number UN1188
Sensitivity Moisture sensitive
Formula Weight 314.165
Concentration or Composition (by Analyte or Components) 5% w/v in 2-methoxyethanol
Physical Form Liquid
Chemical Name or Material Yttrium(III) 2-methoxyethoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.