missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) 2-ethylhexanoate, 99.8% (metals basis)

Product Code. 11378088
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378088 100 g 100g
11388088 5 g 5g
11398088 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11378088 Supplier Thermo Scientific Alfa Aesar Supplier No. 036661.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103470-68-4 | C24H45O6Y | 518.524 g/mol

Yttrium(III) 2-ethylhexanoate has been used for research in nanotechnology based applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103470-68-4
Molecular Formula C24H45O6Y
Molecular Weight (g/mol) 518.524
MDL Number MFCD00070441
InChI Key AGOMHFKGCMKLDA-UHFFFAOYSA-K
Synonym yttrium iii 2-ethylhexanoate, yttrium 2-ethylhexanoate, hexanoic acid,2-ethyl-, yttrium 3+ salt 3:1, yttrium iii 2-ethylhexanoate w/v in n-hexane, acmc-1c5ry, yttrium tri 2-ethylhexanoate, yttrium tris 2-ethylhexanoate, tris 2-ethylhexanoic acid yttrium salt, yttrium 3+ tris 2-ethylhexanoate, yttrium iii 2-ethylhexanoate y
PubChem CID 15607674
IUPAC Name 2-ethylhexanoate;yttrium(3+)
SMILES CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Y+3]

Specifications

Quantity 100 g
Solubility Information Insoluble in water.
Formula Weight 518.16
Percent Purity 99.8%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Yttrium(III) 2-ethylhexanoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.