Learn More
Yttrium(III) 2-ethylhexanoate, 99.8% (metals basis)
Description
Yttrium(III) 2-ethylhexanoate has been used for research in nanotechnology based applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 25 g |
| Solubility Information | Insoluble in water. |
| Formula Weight | 518.16 |
| Percent Purity | 99.8% |
| Physical Form | Powder |
| Assay | (metals basis) |
| Chemical Name or Material | Yttrium(III) 2-ethylhexanoate |
Safety and Handling
- Yttrium(III) 2-ethylhexanoate
- Warning
- Reproductive toxicity Category 2
- H361d-Suspected of damaging the unborn child.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contain: Yttrium(III) 2-ethylhexanoate
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.