missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) 2-ethylhexanoate, 10% w/v in n-hexane

Product Code. p-7058850
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
This item is not returnable. View return policy

Product Code. 11389528

Brand: Thermo Scientific Alfa Aesar 043356.AC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103470-68-4 | C24H45O6Y | 518.524 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103470-68-4
Molecular Formula C24H45O6Y
Molecular Weight (g/mol) 518.524
MDL Number MFCD00070441
InChI Key AGOMHFKGCMKLDA-UHFFFAOYSA-K
Synonym yttrium iii 2-ethylhexanoate, yttrium 2-ethylhexanoate, hexanoic acid,2-ethyl-, yttrium 3+ salt 3:1, yttrium iii 2-ethylhexanoate w/v in n-hexane, acmc-1c5ry, yttrium tri 2-ethylhexanoate, yttrium tris 2-ethylhexanoate, tris 2-ethylhexanoic acid yttrium salt, yttrium 3+ tris 2-ethylhexanoate, yttrium iii 2-ethylhexanoate y
PubChem CID 15607674
IUPAC Name 2-ethylhexanoate;yttrium(3+)
SMILES CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Y+3]

Specifications

Quantity 25 mL
UN Number UN1208
Formula Weight 518.16
Concentration or Composition (by Analyte or Components) 10% w/v in n-hexane
Physical Form Liquid
Chemical Name or Material Yttrium (III) 2-ethylhexanoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.