missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Yttrium(III) 2,4-pentanedionate hydrate, 99.9% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Yttrium(III) 2,4-pentanedionate hydrate acts as a precursor with europium nitrate and used for the preparation of eurobium-doped yttrium orthovanadate nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Powder |
Quantity | 100 g |
Solubility Information | Insoluble in water. |
IUPAC Name | yttrium(3+) tris(2,4-dioxopentan-3-ide) |
Formula Weight | 386.24 (Anhydrous) |
Percent Purity | 99.9% |
Assay | (REO) |
Chemical Name or Material | Yttrium(III) 2,4-pentanedionate hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'