missing translation for 'onlineSavingsMsg'
Learn More

Yttrium Foil, 0.25mm (0.01 in.) thick, 0.999, Alfa Aesar™

Form: Approx. 0.7g/25 x 25mm

Brand:  Alfa Aesar 013960.FI

Code : NEW

Additional Details : CAS Number : 7440-65-5 Weight : 0.01000kg

Quantity 50 x 50mm
Packaging Mylar under Argon

 View more versions of this product

Product Code. 11470380

  • £265.80 / Each


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

Yttrium
7440-65-5
99.90%
Rare Earth Oxides Content
Foil, 0.25mm (0.01 in.) thick
MFCD00011468

Specifications

Yttrium
Rare Earth Oxides Content
Gray foil
Foil, 0.25mm (0.01 in.) thick
MFCD00011468
Mylar under Argon
50 x 50mm
7440-65-5
4.47g/cm3
99.90%
Odorless
Gray
Videos
SDS