missing translation for 'onlineSavingsMsg'
Learn More

Yttrium foil, 0.025mm (0.001in) thick, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number 11400290
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7440-65-5
Y
88.91
MFCD00011468
VWQVUPCCIRVNHF-UHFFFAOYSA-N
yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
23993
CHEBI:33331
yttrium
[Y]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7440-65-5
Y
88.91
MFCD00011468
VWQVUPCCIRVNHF-UHFFFAOYSA-N
yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
23993
CHEBI:33331
yttrium
[Y]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
UN Number UN3178
Quantity 50 x 50 mm
Percent Purity 99.9%
Odor Odorless
Weight ≈0.07g/25x25mm
Form Foil
Assay (REO)
Chemical Name or Material Yttrium foil, 0.025mm (0.001 in.) thick

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'