missing translation for 'onlineSavingsMsg'
Learn More

Yttrium aluminum oxide, 99.9% (REO)

Product Code. 15471707
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15471707 100 g 100g
15461707 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15471707 Supplier Thermo Scientific Alfa Aesar Supplier No. 041491.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rare earth element doped yttrium aluminum oxide is used in solid state lasers | CAS: 12005-21-9

Rare earth element doped yttrium aluminum oxide is used in solid state lasers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12005-21-9
Molecular Formula Y3Al5O12
MDL Number MFCD00213958
Synonym Yttrium aluminum garnet; YAG

Specifications

Melting Point 1760°C
Physical Form Powder
Quantity 100 g
Formula Weight 593.62
Percent Purity 99.9%
Refractive Index 1.833
Density 4.65 g/mL
Assay (REO)
Chemical Name or Material Yttrium aluminum oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.