missing translation for 'onlineSavingsMsg'
Learn More

Yttrium acetate hydrate, 99.9%, (trace metal basis)

Product Code. p-3593546 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10035123

Brand: Thermo Scientific Acros 392210250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 207801-28-3 | C6H9O6Y | 266.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207801-28-3
Molecular Formula C6H9O6Y
Molecular Weight (g/mol) 266.04
MDL Number MFCD00150128
InChI Key JUWHRRAPBUAYTA-UHFFFAOYSA-K
Synonym yttriumacetatetetrahydrate, yttrium acetate-water 1/3/4, yttrium 3+ triacetate tetrahydrate, yttrium 3+ ion triacetate tetrahydrate
PubChem CID 19818017
SMILES [Y+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Color White
Physical Form Crystalline Powder and Lumps
Quantity 25 g
Assay Percent Range 99.90%
Solubility Information Solubility in water: soluble
IUPAC Name yttrium(3+) triacetate
Formula Weight 266.04
Percent Purity 99.9%
Grade Trace Metal Basis
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Yttrium acetate hydrate
Show More Show Less
compliance-icons
Product Identifier
  • Yttrium acetate hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.