missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium ingot, 99.9% (REO), Thermo Scientific Chemicals

£64.70 - £228.00

Chemical Identifiers

CAS 7440-64-4
Molecular Formula Yb
Molecular Weight (g/mol) 173.05
MDL Number MFCD00011286
InChI Key NAWDYIZEMPQZHO-UHFFFAOYSA-N
Synonym unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n
PubChem CID 23992
ChEBI CHEBI:33381
IUPAC Name ytterbium
SMILES [Yb]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11346547
View Documents Promotion Details
Thermo Scientific Alfa Aesar
000407.06
5 g
£64.70
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11336547
View Documents Promotion Details
Thermo Scientific Alfa Aesar
000407.14
25 g
£228.00
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged in air stable poly bottle
Specifications

Chemical Identifiers

7440-64-4
173.05
NAWDYIZEMPQZHO-UHFFFAOYSA-N
23992
ytterbium
Yb
MFCD00011286
unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n
CHEBI:33381
[Yb]

Specifications

7440-64-4
MFCD00011286
NAWDYIZEMPQZHO-UHFFFAOYSA-N
ytterbium
23992
99.9%
Poly Bottle
REO
Yb
unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n
[Yb]
173.05
CHEBI:33381
Odorless
Ingot
Ytterbium ingot
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

missing translation for 'einecsNumber' : 231-173-2

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Packaged in air stable poly bottle

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only