missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) triflate hydrate, 99.9%, (trace metal basis)

Product Code. 10085863 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10085863 5 g Glass bottle 5g
10646871 1 g Glass bottle 1g
10045913 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10085863 Supplier Thermo Scientific Acros Supplier No. 304170050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 252976-51-5 | C3F9O9S3Yb | 620.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 252976-51-5
Molecular Formula C3F9O9S3Yb
Molecular Weight (g/mol) 620.23
MDL Number MFCD06200261
InChI Key AHZJKOKFZJYCLG-UHFFFAOYSA-K
Synonym ytterbium iii trifluoromethanesulfonate hydrate, ytterbium 3+ hydrate tritriflate, ytterbium iii triflate hydrate, ytterbium triflate hydrate, acmc-1aqes, yb otf 3.h2o, ytterbium triflate monohydrate, ytterbium trifluoromethanesulfonate hydrate, ytterbium trifluoromethanesulfonate, hydrate
PubChem CID 22836430
IUPAC Name ytterbium(3+) tritrifluoromethanesulfonate
SMILES [Yb+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Specifications

Melting Point 118.0°C to 122.0°C
Color White
Physical Form Crystalline Powder
Quantity 5 g
Assay Percent Range 99.90%
Linear Formula (CF3SO3)3Yb·xH2O
Solubility Information Solubility in water: soluble
Formula Weight 620.23
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Ytterbium(III) triflate hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.