missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) isopropoxide

Product Code. 11364487
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11364487 5 g 5g
11354487 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11364487 Supplier Thermo Scientific Alfa Aesar Supplier No. 039579.06
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 6742-69-4 | C9H24O3Yb | 353.342 g/mol

Yttrium(III) tris(isopropoxide) is a catalyst for stereoselective conjugate addition reactions, ring-opening polymerization, generation of reactive enolates, allylic substitution and cyano-phosphorylation of enones and synthesis of nanocomposites.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6742-69-4
Molecular Formula C9H24O3Yb
Molecular Weight (g/mol) 353.342
MDL Number MFCD00145532
InChI Key IVGMSSQGEVYWID-UHFFFAOYSA-N
Synonym propan-2-ol; ytterbium, tris isopropanolato ytterbium
PubChem CID 50912003
IUPAC Name propan-2-ol;ytterbium
SMILES CC(C)O.CC(C)O.CC(C)O.[Yb]

Specifications

Melting Point 200°C (decomposition)
Boiling Point 190°C to 200°C (0.2 mmHg)(sublimation)
Odor Odorless
Packaging Ampule
Linear Formula Yb[OCH(CH3)2]3
Quantity 5 g
UN Number UN2925
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water. Soluble in hexane,toluene,and THF.
Formula Weight 350.31
Physical Form Powder
Chemical Name or Material Ytterbium(III) isopropoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.