missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) fluoride, anhydrous, 99.9% (REO)

Product Code. 11334128
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334128 10 g 10g
11344128 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334128 Supplier Thermo Scientific Alfa Aesar Supplier No. 041423.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For vacuum deposition | CAS: 13760-80-0 | F3Yb | 230.04 g/mol

Ytterbium(III) fluoride is used in laboratory reagent, optical glasses, structural ceramics, catalysts, electrical components and photo-optical material. It is also used in used in the preparation and study of fluorinated glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13760-80-0
Molecular Formula F3Yb
Molecular Weight (g/mol) 230.04
MDL Number MFCD00049615
InChI Key XASAPYQVQBKMIN-UHFFFAOYSA-K
Synonym ytterbium 3+ trifluoride, ytterbium 3+ ion trifluoride, acmc-209ceg, ytterbium iii ; fluoride, fluoride, fluoride, yb, fluoride, ytterbium iii fluoride, anhydrous trace rare earth metals basis 10g
PubChem CID 21694467
SMILES [F-].[F-].[F-].[Yb+3]

Specifications

Melting Point 1157°C
Physical Form Pieces
Quantity 10 g
UN Number UN3288
Solubility Information Soluble moderately in strong mineral acids. Insoluble in water.
IUPAC Name ytterbium(3+) trifluoride
Formula Weight 230.04
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 8.2 g/mL
Assay (REO)
Chemical Name or Material Ytterbium(III) fluoride, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.