missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Catalog Number p-4904570
Click to view available options
Quantity:
5 g
25 g
10361-91-8
YbCl3
MFCD00011288
10361-91-8
YbCl3
MFCD00011288

Used as laboratory reagent, optical glasses, structural ceramics, catalysts, electrical components and photo-optical material. Applied with NiCl2 to form a very effective catalyst for the reductive dehalogenation of aryl halides. Ytterbium(III) chloride is a powerful catalyst for the formation of acetals using trimethyl orthoformate and also used to track digestion in animals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 854°C
Color White
Physical Form Beads
Quantity 5 g
Assay Percent Range 99.99% (REO)
Solubility Information Slightly soluble in water.
Formula Weight 279.4
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Ytterbium(III) chloride, ultra dry

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'