missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) bromide hydrate, 99.9% (REO)

Product Code. 11322528
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11322528 10 g 10g
11332528 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322528 Supplier Thermo Scientific Alfa Aesar Supplier No. 014575.09
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15163-03-8

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15163-03-8
MDL Number MFCD00049613
Synonym ytterbium bromide hydrate, acmc-20aj9h, ytterbium 3+ bromide-water 1/3/1

Specifications

Melting Point 300°C
Color White
Physical Form Crystalline
Quantity 10 g
Solubility Information Soluble in water.
Formula Weight 412.77 (Anhydrous)
Percent Purity 99.9%
Sensitivity Very Hygroscopic
Assay (REO)
Chemical Name or Material Ytterbium(III) bromide hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.