missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ YTHDF3 Recombinant Rabbit Monoclonal Antibody (HL2235)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550449
This item is not returnable.
View return policy
Description
YTHDF3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a member of the YTH (YT521-B homology) domain protein family. The YTH domain is common in eukaryotes, is often found in the middle of the protein sequence, and may function in binding to RNA. Alternative splicing results in multiple transcript variants.
Specifications
| YTHDF3 | |
| Recombinant Monoclonal | |
| 2 mg/mL | |
| PBS with no preservative | |
| Q7Z739, Q8BYK6 | |
| YTHDF3 | |
| Recombinant fragment of human YTHDF3. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| HL2235 | |
| Unconjugated | |
| YTHDF3 | |
| 9130022A11Rik; YTH domain family 3; YTH domain family protein 3; YTH domain family, member 3; YTH domain-containing family protein 3; YTH N(6)-methyladenosine RNA binding protein 3; YTH N6-methyladenosine RNA binding protein 3; Ythdf3 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 229096, 253943, 361920 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction