missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ YTHDF3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA526983

Product Code. 13221219

  • £479.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is predicted to react with mouse based on sequence homology.

This gene encodes a member of the YTH (YT521-B homology) domain protein family. The YTH domain is common in eukaryotes, is often found in the middle of the protein sequence, and may function in binding to RNA. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

YTHDF3
Polyclonal
PBS with 0.09% sodium azide; pH 7.4
Q7Z739
YTHDF3
KLH conjugated synthetic peptide between 228-257 amino acids from the central region of human YTHD3.
200 μL
Primary
Human
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
YTHDF3
9130022A11Rik; YTH domain family 3; YTH domain family protein 3; YTH domain family, member 3; YTH domain-containing family protein 3; YTH N(6)-methyladenosine RNA binding protein 3; YTH N6-methyladenosine RNA binding protein 3; Ythdf3
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
253943
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.