missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Specifically recognizes and binds N6-methyladenosine (m6A)-containing RNAs, and regulates mRNA stability. M6A is a modification present at internal sites of mRNAs and some non-coding RNAs and plays a role in the efficiency of mRNA splicing, processing and stability. Acts as a regulator of mRNA stability: binding to m6A-containing mRNAs results in the localization to mRNA decay sites, such as processing bodies (P-bodies), leading to mRNA degradation. Also acts as a promoter of cap-independent mRNA translation following heat shock stress: upon stress, relocalizes to the nucleus and specifically binds mRNAs with some m6A methylation mark at their 5′-UTR, protecting demethylation of mRNAs by FTO, thereby promoting cap-independent mRNA translation.
Specifications
Specifications
| Antigen | YTHDF2 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | YTHDF2 |
| Gene Accession No. | Q9Y5A9 |
| Gene Alias | CLL associated antigen KW 14, HGRG8, NY REN 2, YTH domain family protein 2, YTH domain family, member 2, YTHDF2 |
| Gene Symbols | YTHDF2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?