missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YTHDF2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-68614
This item is not returnable.
View return policy
Description
YTHDF2 Polyclonal antibody specifically detects YTHDF2 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| YTHDF2 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
| CLL-associated antigen KW-14, HGRG8YTH domain family 2, High-glucose-regulated protein 8, NY-REN-2,9430020E02Rik, Renal carcinoma antigen NY-REN-2, YTH domain family protein 2, YTH domain family, member 2 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: TQPLPPPPPQPAQLSVQQQAAQPTRWVAPRNRGSGFGHNGVDGNGVGQSQAGSGSTPSEPHPVLEKLRSIN | |
| 100 μg | |
| Cell Biology | |
| 51441 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction