missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YTHDF1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26787-1-AP-150UL
This item is not returnable.
View return policy
Description
Specifically recognizes and binds N6-methyladenosine (m6A)-containing RNAs. M6A is a modification present at internal sites of mRNAs and some non-coding RNAs and plays a role in the efficiency of mRNA splicing, processing and stability. Ota T., Nat. Genet. 36:40-45(2004). Deloukas P., Nature 414:865-871(2001). The MGC Project Team, Genome Res. 14:2121-2127(2004).Specifications
| YTHDF1 | |
| Polyclonal | |
| Unconjugated | |
| YTHDF1 | |
| C20orf21, DACA 1, YTH domain family protein 1, YTH domain family, member 1, YTHDF1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54915 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9BYJ9 | |
| YTHDF1 | |
| YTHDF1 Fusion Protein Ag25347 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction