Learn More
YTHDF1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26787-1-AP-20UL
 View more versions of this product
Product Code. 16805155
  • £135.00 / 20µL
Description
Specifically recognizes and binds N6-methyladenosine (m6A)-containing RNAs. M6A is a modification present at internal sites of mRNAs and some non-coding RNAs and plays a role in the efficiency of mRNA splicing, processing and stability. Ota T., Nat. Genet. 36:40-45(2004). Deloukas P., Nature 414:865-871(2001). The MGC Project Team, Genome Res. 14:2121-2127(2004).
Specifications
Specifications
YTHDF1
Polyclonal
Unconjugated
YTHDF1
C20orf21, DACA 1, YTH domain family protein 1, YTH domain family, member 1, YTHDF1
Rabbit
Antigen Affinity Chromatography
RUO
54915
-20°C
Liquid
Immunoprecipitation, Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BYJ9
YTHDF1
YTHDF1 Fusion Protein Ag25347
20 μL