missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ YTHDF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570276
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 86%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
Specifically recognizes and binds N6-methyladenosine (m6A)-containing mRNAs, and promotes mRNA translation efficiency (PubMed:24284625, PubMed:26046440, PubMed:26318451). M6A is a modification present at internal sites of mRNAs and some non-coding RNAs and plays a role in the efficiency of mRNA splicing, processing and stability (PubMed:24284625). Acts as a regulator of mRNA translation efficiency: promotes ribosome loading to m6A-containing mRNAs and interacts with translation initiation factors eIF3 (EIF3A or EIF3B) to facilitate translation initiation (PubMed:26046440). Required to facilitate learning and memory formation in the hippocampus by enhancing protein synthesis upon neuronal stimulation: in response to neuronal stimulation, binds to m6A-containing neuronal mRNAs, promoting their translation, thereby contributing to learning and memory. Acts as a regulator of axon guidance by binding to m6A-containing ROBO3 transcripts, thereby promoting their translation. Acts as a negative regulator of antigen cross-presentation in myeloid dendritic cells. Acts by binding and promoting translation of m6A-containing transcripts encoding proteins involved in lysosomal degradation and phagosome maturation, leading to increased antigen degradation in myeloid dendritic cells. In the context of tumorigenesis, negative regulation of antigen cross-presentation limits the anti-tumor response by reducing efficiency of tumor-antigen cross-presentation. [UniProt].
Specifications
| YTHDF1 | |
| Polyclonal | |
| Unconjugated | |
| YTHDF1 | |
| 2210410K23Rik; 8030473O16; C20orf21; DACA-1; DACA-1 homolog; Dermatomyositis associated with cancer putative autoantigen 1; dermatomyositis associated with cancer putative autoantigen 1 homolog; fb49c02; wu:fb49c02; wu:fi19h06; wu:fj87f09; wu:fp06c11; YTH domain family 1; YTH domain family protein 1; YTH domain family, member 1; YTH domain-containing family protein 1; YTH N(6)-methyladenosine RNA binding protein 1; YTH N6-methyladenosine RNA binding protein 1; Ythdf1; zgc:55657 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 296467 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| YTHDF1 | |
| synthetic peptide directed towards the N-terminal region of rat YTHDF1. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction