Learn More
Invitrogen™ YTHDC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144005
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human Daudi whole cell, human Hela whole cell, human 293T whole cell, rat C6 whole cell, mouse NIH/3T3 whole cell. ICC/IF: SiHa cell. Flow: MCF-7 cell, SiHa cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Specifically recognizes and binds N6-methyladenosine (m6A)-containing RNAs. M6A is a modification present at internal sites of mRNAs and some non-coding RNAs and plays a role in the efficiency of mRNA splicing, processing and stability.
Specifications
| YTHDC2 | |
| Polyclonal | |
| Unconjugated | |
| YTHDC2 | |
| 3010002F02Rik; 3'-5' RNA helicase YTHDC2; BC037178; CAHL; CsA-associated helicase-like protein; DKFZp564A186; FLJ10053; FLJ2194; hYTHDC2; Keen to exit meiosis leaving testes under-populated protein; Ketu; mYTHDC2; probable ATP-dependent RNA helicase YTHDC2; YTH domain containing 2; YTH domain-containing protein 2; YTH domain-containing protein C2; Ythdc2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 240255, 307446, 64848 | |
| -20°C | |
| Lyophilized |
| ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| B2RR83, Q9H6S0 | |
| YTHDC2 | |
| E.coli-derived human YTHDC2 recombinant protein (Position: K105-K1231). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.