missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YTHD1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94472-0.02ml
This item is not returnable.
View return policy
Description
YTHD1 Polyclonal antibody specifically detects YTHD1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| YTHD1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| C20orf21, DACA-1, Dermatomyositis associated with cancer putative autoantigen 1, FLJ20391, YTH domain family 1, YTH domain family protein 1, YTH domain family, member 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 280-390 of human YTHD1 (NP_060268.2). VPKAPVPQQAPSPQAAPQPQQVAQPLPAQPPALAQPQYQSPQQPPQTRWVAPRNRNAAFGQSGGAGSDSNSPGNVQPNSAPSVESHPVLEKLKAAHSYNPKEFEWNLKSGR | |
| 0.02 mL | |
| Cell Biology | |
| 54915 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction