missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ YT-Broth Agar (Capsules)
Shop All MP Biomedicals Products
Click to view available options
:
5kg
Specifications
Specifications
| Certifications/Compliance | Molecular Biology Certified™ |
| Content And Storage | Store at room temperature. |
| Description | YT-broth agar |
| Format | Capsules |
| Formulation | Per Liter: 8 g Tryptone-B; 5 g Yeast Extract-B; 5 g NaCl; 15 g Agar-B |
| For Use With (Application) | Bacterial culture |
| Quantity | 5 kg |
| Specimen Type | E. coli |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction