Learn More
YSI™ Probe Service Kits for YSI Dissolved Oxygen Probes
Product Code. 12704576
Click to view available options
Type:
High-sensitivity Membrane Kit
Standard Membrane Kit
Quantity:
10 x 150 Membranes
30 Membranes/Pk.
Unit Size:
Each
Product Code. 12704576
Brand: YSI™ 098095
Only null left
Add to basket
Add to basket
For replenishing oxygen sensor on YSI Dissolved Oxygen Probes
  • Membrane Kits include 150 standard, 1mil PTFE membranes or 150 high-sensitivity, 0.5mil PTFE membranes, in 10 packs of 50
  • Membrane/KCI Kits include 30 standard or high-sensitivity membranes, 1 oz. (30mL) dropper bottle with KCI crystals for adding distilled water and a probe contour tool
  • For use with probes 13-299-43 (Model 5739), 13-298-80 (Model 5750), 15-176-50 (Model 5718), 13-298-16 (Model 5300 Biological Oxygen Monitor), and 13-298-56, -57, or -62 (Model 55 Handheld Dissolved Oxygen Meters)
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Replenishing Oxygen Sensor on YSI™ DO Probe
Type High-sensitivity Membrane Kit
Includes 30 High sensitivity membrane
Probe Type DO
Quantity 30 Membranes/Pk.