Learn More
YSI™ Probe Service Kits for YSI Dissolved Oxygen Probes
Product Code. 12794566
Change view
Click to view available options
Type:
High-sensitivity Membrane Kit
Standard Membrane Kit
Quantity:
10 x 150 Membranes
30 Membranes/Pk.
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Quantity unitSize
12794566 Standard Membrane Kit 30 Membranes/Pk. -
12724576 High-sensitivity Membrane Kit 10 x 150 Membranes Each
12704576 High-sensitivity Membrane Kit 30 Membranes/Pk. -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12794566 Supplier YSI™ Supplier No. 098094
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For replenishing oxygen sensor on YSI Dissolved Oxygen Probes
  • Membrane Kits include 150 standard, 1mil PTFE membranes or 150 high-sensitivity, 0.5mil PTFE membranes, in 10 packs of 50
  • Membrane/KCI Kits include 30 standard or high-sensitivity membranes, 1 oz. (30mL) dropper bottle with KCI crystals for adding distilled water and a probe contour tool
  • For use with probes 13-299-43 (Model 5739), 13-298-80 (Model 5750), 15-176-50 (Model 5718), 13-298-16 (Model 5300 Biological Oxygen Monitor), and 13-298-56, -57, or -62 (Model 55 Handheld Dissolved Oxygen Meters)
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 5718, 5739, 5750, 5300 and Rapid Pulse Probes
Type Standard Membrane Kit
Includes Bottle of electrolyte, 2 O-rings, 30 1mL FEP Fluoropolymer Film Membranes
Probe Type DO
Quantity 30 Membranes/Pk.
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.