missing translation for 'onlineSavingsMsg'
Learn More

YSI™ pH/ORP Dual Probes for Pro Series Portable Meters

Product Code. 12913507
Change view
Click to view available options
Type:
pH/ORP probe
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
12913507 pH/ORP probe Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12913507 Supplier YSI™ Supplier No. 605103

Please to purchase this item. Need a web account? Register with us today!

Edge

For use with Professional Plus Multiparameter Handheld Instrument.

  • Easy user-replaceble sensor
  • Measure pH and ORP or redox quickly and easily
  • Requires cable/probe assembly
TRUSTED_SUSTAINABILITY

Specifications

Type pH/ORP probe
For Use With (Equipment) Professional Plus Multiparameter Handheld Instrument
Probe pH/ORP dual probe
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.