missing translation for 'onlineSavingsMsg'
Learn More

YSI™ Nitrate Calibration Solutions

Product Code. 12746955
Click to view available options
Concentration:
1 mg/L
10 mg/L
100 mg/L
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12746955

Brand: YSI™ 003887

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extremely accurate calibration solutions to calibrate field instruments equipped with a nitrate sensor

  • Plastic bottle (500mL)
  • Concentration options: 1m/L, 10mg/L, 100mg/L

Expiration Dates:
Unopended: 1 year
Opened: 6 months

TRUSTED_SUSTAINABILITY

Specifications

Concentration 100 mg/L
Quantity 1 Ea.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.