Learn More
YSI™ Cap Membrane Kits
Brand: YSI™
Code : 03
Additional Details : CAS Number : 7447-40-7 Weight : 0.05452kg
Description
Includes six cap membranes, a 1 oz. (30mL) bottle of KCl solution and one sanding disk. For use with 13-398-87, -88, and -89 (Model 5239 Field Oxygen/Temperature Probes); 13-299-76 (Model 5905 BOD Probe); and 13-298-24 through-27 (Model 85 DO/Conductivity Meters).
Specifications
Cap Membrane Kits | |
Model 5239 Field Oxygen/Temperature Probes; Model 5905 BOD Probe; and Model 85 DO/Conductivity Meters | |
Six cap membranes, a 1 oz. (30 mL) bottle of KCl solution and one sanding disk |