Learn More
YSI™ Bar Code Scanner for Models 5000 and 5100 Dissolved Oxygen Meters
Product Code. 12744566
Product Code. 12744566
Brand: YSI™ 051060
Eliminates need to manually type or write bottle numbers
Bar Code Scanner. For use with Model 5100. One swipe of scanner records the BOD bottle number, DO concentration, temperature and time of day. Eliminates need to manually type or write bottle numbers.
TRUSTED_SUSTAINABILITY
Specifications
Type Bar Code Scanner
For Use With (Application) One swipe of scanner records the BOD bottle number, DO concentration, temperature and time of day