Learn More
YSI™ 6052030 Pro Series DO/Conductivity Cables
Cables available in 4m, 10, 20, 30m lengths
Brand:  YSI™ 6052030-30
 View more versions of this product
Product Code. 11927127
  • £885.00 / Each
Estimated Shipment: 25-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
The 6052030 cables ONLY work with the YSI Pro Plus handheld
  • Measure dissolved oxygen, conductivity, salinity, specific conductance, total dissolved solids and temperature
  • Conductivity sensor is built into the cable
  • Automatic salinity compensation for DO readings
  • Cables do NOT come with the DO sensor
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Conductivity, DO, Temp
Built-in conductivity sensor and temperature/DO sensor port
1181.1 in.
30 m