Learn More
YSI™ 6052030 Pro Series DO/Conductivity Cables
Click to view available options
Length (English):
157.4 in.
393.7 in.
787.4 in.
1181.1 in.
Unit Size:
Each
Description
The 6052030 cables ONLY work with the YSI Pro Plus handheld
- Measure dissolved oxygen, conductivity, salinity, specific conductance, total dissolved solids and temperature
- Conductivity sensor is built into the cable
- Automatic salinity compensation for DO readings
- Cables do NOT come with the DO sensor
Specifications
Specifications
| Includes | Built-in conductivity sensor and temperature/DO sensor port |
| Length (English) | 787.4 in. |
| Length (Metric) | 20 m |