missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YS-49, MedChemExpress
Description
YS-49 is a PI3K/Akt (a downstream target of RhoA) activator, to reduce RhoA/PTEN activation in the 3-methylcholanthrene-treated cells. YS-49 inhibits angiotensin II (Ang II)-stimulated proliferation of VSMCs via induction of heme oxygenase (HO)-1. YS-49 is also an isoquinoline compound alkaloid, has a strong positive inotropic action through activation of cardiac β-adrenoceptors.
Specifications
Specifications
| Chemical Name or Material | YS-49 |
| CAS | 132836-42-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C20H20BrNO2 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 100 mg/mL (258.88 mM; Need ultrasonic) ∣H2O : 10 mg/mL (25.89 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | OC1=CC2=C(C(CC3=C4C=CC=CC4=CC=C3)NCC2)C=C1O.[H]Br |
| Molecular Weight (g/mol) | 386.28 |
| Formula Weight | 386.28 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?